4-Methylthiophene-3-boronic acid pinacol ester

Supplier: Apollo Scientific

Synonyms: 4-Methylthiophene-3-boronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-(4-methylthiophen-3-yl)-1,3,2-dioxaborolane

OR310162-1G OR310162-500MG
APOSOR310162-1GEA 439 GBP
APOSOR310162-1G APOSOR310162-500MG
4-Methylthiophene-3-boronic acid pinacol ester
4-Methylthiophene-3-boronic acid pinacol ester
Formula: C₁₁H₁₇BO₂S
MW: 224,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09265133
CAS Number: 476620-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR