2-Chloro-5-iodo-6-methylpyridine

Supplier: Apollo Scientific

Synonyms: 6-Chloro-3-iodo-2-picoline, 6-Chloro-3-iodo-2-methylpyridine, 2-Chloro-5-iodo-6-picoline

OR310156-1G OR310156-500MG
APOSOR310156-1GEA 439 GBP
APOSOR310156-1G APOSOR310156-500MG
2-Chloro-5-iodo-6-methylpyridine
2-Chloro-5-iodo-6-methylpyridine
Formula: C₆H₅ClIN
MW: 253,47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09037461
CAS Number: 249291-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR