2-Amino-3-iodo-5-methylpyridine
Supplier: Apollo Scientific
Synonyms:
3-Iodo-5-methyl-2-pyridylamine, 2-Amino-3-iodo-5-picoline, 3-Iodo-5-methylpyridin-2-amine
OR310151-1G
OR310151-500MG
APOSOR310151-1GEA
439
GBP
APOSOR310151-1G
APOSOR310151-500MG
2-Amino-3-iodo-5-methylpyridine
2-Amino-3-iodo-5-methylpyridine
Formula:
C₆H₇IN₂ MW: 234,04 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09037445 CAS Number: 211308-79-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...