3-Bromo-1H-indole-2-carbonitrile

Supplier: Apollo Scientific

OR310141-1G OR310141-500MG
APOSOR310141-1GEA 439 GBP
APOSOR310141-1G APOSOR310141-500MG
3-Bromo-1H-indole-2-carbonitrile
3-Bromo-1H-indole-2-carbonitrile
Formula: C₉H₅BrN₂
Melting Pt: 176…178 °C
Storage Temperature: Ambient
MDL Number: MFCD22126115
CAS Number: 1373348-93-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR