3-Bromo-2-ethoxypyridine
Supplier: Apollo Scientific
OR310089-1G
OR310089-500MG
APOSOR310089-1GEA
328
GBP
APOSOR310089-1G
APOSOR310089-500MG
3-Bromo-2-ethoxypyridine
3-Bromo-2-ethoxypyridine
Formula:
C₇H₈BrNO MW: 202,05 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00234309 CAS Number: 57883-25-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...