N,N-Diethylethylenediamine 97%

Supplier: Apollo Scientific
Danger

Synonyms: 2-(Diethylamino)ethylamine, 2-Aminoethyldiethylamine

OR31005-1KG OR31005-500G
APOSOR31005-1KGEA 197 GBP
APOSOR31005-1KG APOSOR31005-500G
N,N-Diethylethylenediamine 97%
N,N-Diethylethylenediamine
Formula: C₆H₁₆N₂
MW: 116,21 g/mol
Boiling Pt: 145…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00008176
CAS Number: 100-36-7
UN: 2685
ADR: 8+3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR