4-tert-Butylpyrimidin-2-amine 95%

Supplier: Apollo Scientific
Warning

OR310038-1G OR310038-250MG
APOSOR310038-1GEA 499 GBP
APOSOR310038-1G APOSOR310038-250MG
4-tert-Butylpyrimidin-2-amine 95%
4-tert-Butylpyrimidin-2-amine
Formula: C₈H₁₃N₃
Melting Pt: 126…128 °C
Storage Temperature: Ambient
MDL Number: MFCD01646229
CAS Number: 17321-94-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR