4-(Dibenzylamino)benzeneboronic acid
Supplier: Apollo Scientific
OR310016-1G
OR310016-500MG
APOSOR310016-1GEA
223
GBP
APOSOR310016-1G
APOSOR310016-500MG
4-(Dibenzylamino)benzeneboronic acid
4-(Dibenzylamino)benzeneboronic acid
Formula:
C₂₀H₂₀BNO₂ MW: 317,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06798225 CAS Number: 159191-44-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...