2-Benzyloxy-5-bromopyridine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(Benzyloxy)-5-bromopyridine
OR310014-1G
OR310014-500MG
APOSOR310014-1GEA
223
GBP
APOSOR310014-1G
APOSOR310014-500MG
2-Benzyloxy-5-bromopyridine
2-Benzyloxy-5-bromopyridine
Formula:
C₁₂H₁₀BrNO MW: 264,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09037462 CAS Number: 83664-33-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...