3-Amino-1H-pyrazole
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3-Pyrazolamine, 3-AP, 1H-Pyrazol-3-amine, 3-Aminopyrazole
OR30957-25G
OR30957-5G
APOSOR30957-25GEA
34.1
GBP
APOSOR30957-25G
APOSOR30957-5G
3-Amino-1H-pyrazole
3-Amino-1H-pyrazole
Formula:
C₃H₅N₃ Boiling Pt: 218 °C/122mmHg Melting Pt: 34…39 °C Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005236 CAS Number: 1820-80-0 EINECS: 217-338-1 UN: 3259 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...