Ethyl-8-bromo-4-chloroquinoline-3-carboxylate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 8-bromo-4-chloroquinoline-3-carboxylate, 8-Bromo-4-chloroquinoline-3-carboxylic acid ethyl ester

OR309352-1G OR309352-5G
APOSOR309352-1GEA 59.1 GBP
APOSOR309352-1G APOSOR309352-5G
Ethyl-8-bromo-4-chloroquinoline-3-carboxylate
Ethyl-8-bromo-4-chloroquinoline-3-carboxylate
Formula: C₁₂H₉BrClNO₂
MW: 314,563 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173385
CAS Number: 206258-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR