7-Bromoquinolin-4-ol

Supplier: Apollo Scientific
Warning

Synonyms: 7-Bromo-4-hydroxyquinoline

OR309241-1G OR309241-5G
APOSOR309241-1GEA 84 GBP
APOSOR309241-1G APOSOR309241-5G
7-Bromoquinolin-4-ol
7-Bromoquinolin-4-ol
Formula: C₉H₆BrNO
MW: 224,0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659037
CAS Number: 82121-06-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR