6-Bromo-4-hydroxyquinoline-3-carboxylic acid
Supplier: Apollo Scientific
OR309205-1G
OR309205-5G
APOSOR309205-1GEA
57.7
GBP
APOSOR309205-1G
APOSOR309205-5G
6-Bromo-4-hydroxyquinoline-3-carboxylic acid
6-Bromo-4-hydroxyquinoline-3-carboxylic acid
Formula:
C₁₀H₆BrNO₃ MW: 268,06 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01569278 CAS Number: 98948-95-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...