Ethyl-1-oxaspiro[2.5]octane-6-carboxylate 95+%

Supplier: Apollo Scientific

OR309119-1G OR309119-500MG OR309119-5G
APOSOR309119-1GEA 263 GBP
APOSOR309119-1G APOSOR309119-500MG APOSOR309119-5G
Ethyl-1-oxaspiro[2.5]octane-6-carboxylate 95+%
Ethyl-1-oxaspiro[2.5]octane-6-carboxylate
Formula: C₁₀H₁₆O₃
MW: 184,24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12498695
CAS Number: 171361-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR