1-Benzylazetidin-3-ol 98%

Supplier: Apollo Scientific
Warning

OR309026-100G OR309026-25G OR309026-5G
APOSOR309026-100GEA 429 GBP
APOSOR309026-100G APOSOR309026-25G APOSOR309026-5G
1-Benzylazetidin-3-ol 98%
1-Benzylazetidin-3-ol
Formula: C₁₀H₁₃NO
Storage Temperature: Ambient
MDL Number: MFCD00963607
CAS Number: 54881-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR