5-(Aminomethyl)-1H-indazole

Supplier: Apollo Scientific
Warning

OR30830-1G OR30830-250MG
APOSOR30830-1GEA 307 GBP
APOSOR30830-1G APOSOR30830-250MG
5-(Aminomethyl)-1H-indazole
5-(Aminomethyl)-1H-indazole
Formula: C₈H₉N₃
Storage Temperature: Refrigerator
MDL Number: MFCD09028157
CAS Number: 267413-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR