tert-Butyl-7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate

Supplier: Apollo Scientific

Synonyms: tert-Butyl 7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate

OR308133-1G OR308133-5G
APOSOR308133-1GEA 46 GBP
APOSOR308133-1G APOSOR308133-5G
tert-Butyl-7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate
tert-Butyl-7-oxa-3-azabicyclo[4.1.0]heptane-3-carboxylate
Formula: C₁₀H₁₇NO₃
MW: 199,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12407050
CAS Number: 161157-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR