7-Azaindole-5-boronic acid

Supplier: Apollo Scientific

Synonyms: (1H-Pyrrolo[2,3-b]pyridin-5-yl)boronic acid, {1H-pyrrolo[2,3-b]pyridin-5-yl}boronic acid

OR308041-1G OR308041-250MG
APOSOR308041-1GEA 118 GBP
APOSOR308041-1G APOSOR308041-250MG
7-Azaindole-5-boronic acid
7-Azaindole-5-boronic acid
Formula: C₇H₇BN₂O₂
MW: 161,96 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08234881
CAS Number: 944059-24-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR