2-Chloro-6-methoxy-3-methylquinoline

Supplier: Apollo Scientific

OR307923-1G OR307923-5G
APOSOR307923-1GEA 264 GBP
APOSOR307923-1G APOSOR307923-5G
2-Chloro-6-methoxy-3-methylquinoline
2-Chloro-6-methoxy-3-methylquinoline
Formula: C₁₁H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD02179792
CAS Number: 123990-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR