Bis(4-bromophenyl)ether 99%
Supplier: Apollo Scientific
Synonyms:
4,4'-Dibromodiphenyl Ether, 4,4'-Oxybis(bromobenzene), 4 4'-OXYBIS(BROMOBENZENE)
OR3076-100G
OR3076-25G
APOSOR3076-100GEA
0
GBP
APOSOR3076-100G
APOSOR3076-25G
Bis(4-bromophenyl)ether 99%
Bis(4-bromophenyl)ether
Formula:
C₁₂H₈Br₂O MW: 328,00 g/mol Boiling Pt: 338…340 °C Melting Pt: 61…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000095 CAS Number: 2050-47-7 EINECS: 218-090-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...