(S)-2-Amino-3-methoxypropanoic acid hydrochloride 95+%
Supplier: Apollo Scientific
OR307094-1G
OR307094-5G
APOSOR307094-1GEA
69.5
GBP
APOSOR307094-1G
APOSOR307094-5G
(S)-2-Amino-3-methoxypropanoic acid hydrochloride 95+%
(S)-2-Amino-3-methoxypropanoic acid hydrochloride
Formula:
C₄H₁₀ClNO₃ MW: 155,58 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03840366 CAS Number: 336100-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...