N-(α)-Boc-DL-2,3-diaminopropionic acid 95+%

Supplier: Apollo Scientific

Synonyms: 3-Amino-2-((tert-butoxycarbonyl)amino)propanoic acid, 3-Amino-N-(tert-butoxycarbonyl)alanine, Boc-DL-α-3-NH₂-Ala-OH

OR307059-10G OR307059-5G
APOSOR307059-10GEA 70.9 GBP
APOSOR307059-10G APOSOR307059-5G
N-(α)-Boc-DL-2,3-diaminopropionic acid 95+%
N-(alpha)-Boc-DL-2,3-diaminopropionic acid
Formula: C₈H₁₆N₂O₄
MW: 204,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00736752
CAS Number: 159002-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR