4-Chloro-6-iodoquinazoline 95+%

Supplier: Apollo Scientific
Warning

OR30701-10G OR30701-5G
APOSOR30701-10GEA 65.6 GBP
APOSOR30701-10G APOSOR30701-5G
4-Chloro-6-iodoquinazoline 95+%
4-Chloro-6-iodoquinazoline
Formula: C₈H₄ClIN₂
MW: 290,4900 g/mol
Melting Pt: 177 °C
Storage Temperature: Ambient
MDL Number: MFCD01862193
CAS Number: 98556-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR