(4-Methoxypyridin-2-yl)methanamine 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
2-(Aminomethyl)-4-methoxypyridine
OR306389-1G
OR306389-250MG
OR306389-5G
APOSOR306389-1GEA
231
GBP
APOSOR306389-1G
APOSOR306389-250MG
APOSOR306389-5G
(4-Methoxypyridin-2-yl)methanamine 98%
(4-Methoxypyridin-2-yl)methanamine
Formula:
C₇H₁₀N₂O MW: 138,1700 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11111886 CAS Number: 194658-14-5 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...