Ethyl-4-(bromomethyl)benzoate 96%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Ethyl 4-(bromomethyl)benzoate
OR30551-100G
OR30551-25G
OR30551-5G
APOSOR30551-100GEA
474
GBP
APOSOR30551-100G
APOSOR30551-25G
APOSOR30551-5G
Ethyl-4-(bromomethyl)benzoate 96%
Ethyl 4-(bromomethyl)benzoate
Formula:
C₁₀H₁₁BrO₂ MW: 243,1 g/mol Boiling Pt: 145 °C/5mmHg Melting Pt: 28…33 °C Storage Temperature: Ambient |
MDL Number:
MFCD03791307 CAS Number: 26496-94-6 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...