2-Chloropyrimidine-4-carbonitrile
Supplier: Apollo Scientific
|
Warning
|
OR30543-1G
OR30543-5G
APOSOR30543-1GEA
18.5
GBP
APOSOR30543-1G
APOSOR30543-5G
2-Chloropyrimidine-4-carbonitrile
2-Chloropyrimidine-4-carbonitrile
Formula:
C₅H₂ClN₃ MW: 139,5400 g/mol Melting Pt: 53…55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00234989 CAS Number: 75833-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...