(3-Bromo-4-methoxyphenyl)acetic acid

Supplier: Apollo Scientific
Warning

Synonyms: 3-Bromo-4-methoxyphenylacetic acid, 2-(3-Bromo-4-methoxyphenyl)acetic acid

OR3045-25G OR3045-5G
APOSOR3045-25GEA 128 GBP
APOSOR3045-25G APOSOR3045-5G
(3-Bromo-4-methoxyphenyl)acetic acid
(3-Bromo-4-methoxyphenyl)acetic acid
Formula: C₉H₉BrO₃
MW: 245,07 g/mol
Melting Pt: 115…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00017538
CAS Number: 774-81-2
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR