5-Bromo-2-furaldehyde

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromofuran-2-carbaldehyde, 5-Bromofurfural

OR30421-5G OR30421-100G OR30421-25G
APOSOR30421-5GEA 60.3 GBP
APOSOR30421-5G APOSOR30421-100G APOSOR30421-25G
5-Bromo-2-furaldehyde
5-Bromo-2-furaldehyde
Formula: C₅H₃BrO₂
MW: 174,98 g/mol
Boiling Pt: 112 °C/16mmHg
Melting Pt: 82…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00159501
CAS Number: 1899-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR