(R)-2-Amino-1,1,2-triphenylethanol

Supplier: Apollo Scientific

OR304029-100MG OR304029-1G OR304029-250MG
APOSOR304029-100MGEA 109 GBP
APOSOR304029-100MG APOSOR304029-1G APOSOR304029-250MG
(R)-2-Amino-1,1,2-triphenylethanol
(R)-2-Amino-1,1,2-triphenylethanol
Formula: C₂₀H₁₉NO
MW: 289,3700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03093538
CAS Number: 79868-79-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR