2-Bromo-5-methoxy-4-methylbenzoic acid

Supplier: Apollo Scientific

OR303909-1G OR303909-5G OR303909-10G
APOSOR303909-1GEA 129 GBP
APOSOR303909-1G APOSOR303909-5G APOSOR303909-10G
2-Bromo-5-methoxy-4-methylbenzoic acid
2-Bromo-5-methoxy-4-methylbenzoic acid
Formula: C₉H₉BrO₃
Melting Pt: 189…191 °C
Storage Temperature: Ambient
MDL Number: MFCD20921580
CAS Number: 61809-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR