4-Hydroxy-5-methoxy-2-pyridinecarbaldehyde
Supplier: Apollo Scientific
OR303865-1G
OR303865-500MG
APOSOR303865-1GEA
704
GBP
APOSOR303865-1G
APOSOR303865-500MG
4-Hydroxy-5-methoxy-2-pyridinecarbaldehyde
4-Hydroxy-5-methoxy-2-pyridinecarbaldehyde
Formula:
C₇H₇NO₃ Melting Pt: 173…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD12922688 CAS Number: 204847-72-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...