6-Bromo-5-methoxy-2-pyridinecarbaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
6-Bromo-5-methoxypicolinaldehyde
OR303822-1G
OR303822-500MG
APOSOR303822-1GEA
575
GBP
APOSOR303822-1G
APOSOR303822-500MG
6-Bromo-5-methoxy-2-pyridinecarbaldehyde
6-Bromo-5-methoxy-2-pyridinecarbaldehyde
Formula:
C₇H₆BrNO₂ MW: 216,04 g/mol Melting Pt: 122…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD13193086 CAS Number: 329217-74-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...