3-Nitro-2-picoline

Supplier: Apollo Scientific
Warning

Synonyms: 2-Methyl-3-nitropyridine

OR303658-100G OR303658-25G OR303658-5G
APOSOR303658-100GEA 289 GBP
APOSOR303658-100G APOSOR303658-25G APOSOR303658-5G
3-Nitro-2-picoline
3-Nitro-2-picoline
Formula: C₆H₆N₂O₂
MW: 138,1240 g/mol
Melting Pt: 32…33 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04114137
CAS Number: 18699-87-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR