2-(2-Chloroethyl)pyridine

Supplier: Apollo Scientific
Warning

OR303592-1G OR303592-5G
APOSOR303592-1GEA 165 GBP
APOSOR303592-1G APOSOR303592-5G
2-(2-Chloroethyl)pyridine
2-(2-Chloroethyl)pyridine
Formula: C₇H₈ClN
Storage Temperature: Freezer
MDL Number: MFCD06656799
CAS Number: 16927-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR