2-[4-(3-Nitro-2-pyridinyl)piperazino]-1-ethanol

Supplier: Apollo Scientific

OR303590-1G OR303590-500MG OR303590-5G
APOSOR303590-1GEA 148 GBP
APOSOR303590-1G APOSOR303590-500MG APOSOR303590-5G
2-[4-(3-Nitro-2-pyridinyl)piperazino]-1-ethanol
2-[4-(3-Nitro-2-pyridinyl)piperazino]-1-ethanol
Formula: C₁₁H₁₆N₄O₃
Storage Temperature: Ambient
MDL Number: MFCD00278397
CAS Number: 215434-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR