4,4'-Biphenyldiboronic acid bis(pinacol)ester

Supplier: Apollo Scientific

Synonyms: 4,4,5,5-Tetramethyl-2-{4-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]phenyl}-1,3,2-dioxaborolane, 4,4'-Bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,1'-biphenyl

OR303502-10G OR303502-1G OR303502-5G
APOSOR303502-10GEA 127 GBP
APOSOR303502-10G APOSOR303502-1G APOSOR303502-5G
4,4'-Biphenyldiboronic acid bis(pinacol)ester
4,4'-Biphenyldiboronic acid bis(pinacol) ester
Formula: C₂₄H₃₂B₂O₄
MW: 406,14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669545
CAS Number: 207611-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR