Methyl-5-bromothiophene-2-carboxylate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Bromo-thiophene-2-carboxylic acid methyl ester, Methyl 5-bromothiophene-2-carboxylate
OR303229-1G
OR303229-500MG
APOSOR303229-1GEA
106
GBP
APOSOR303229-1G
APOSOR303229-500MG
Methyl-5-bromothiophene-2-carboxylate
Methyl-5-bromothiophene-2-carboxylate
Formula:
C₆H₅BrO₂S MW: 221,0700 g/mol Boiling Pt: 251,868 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03233845 CAS Number: 62224-19-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...