tert-Butyl Hydrogen tetradecanedioate

Supplier: Apollo Scientific
Warning

Synonyms: 14-(tert-Butoxy)-14-oxotetradecanoic acid, Tetradecanedioic acid Mono-tert-butyl ester

OR303208-1G OR303208-5G
APOSOR303208-1GEA 436 GBP
APOSOR303208-1G APOSOR303208-5G
tert-Butyl Hydrogen tetradecanedioate
tert-Butyl Hydrogen tetradecanedioate
Formula: C₁₈H₃₄O₄
MW: 314,47 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23136040
CAS Number: 234082-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR