3-(3-Nitro-4-tetrahydro-1H-pyrrol-1-ylphenyl)acrylic acid

Supplier: Apollo Scientific

OR30320-10G OR30320-1G OR30320-5G
APOSOR30320-10GEA 570 GBP
APOSOR30320-10G APOSOR30320-1G APOSOR30320-5G
3-(3-Nitro-4-tetrahydro-1H-pyrrol-1-ylphenyl)acrylic acid
3-(3-Nitro-4-tetrahydro-1H-pyrrol-1-ylphenyl)acrylic acid
Formula: C₁₃H₁₄N₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00173707
CAS Number: 175278-41-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR