tert-Butyl-3-hydroxypent-4-enoate

Supplier: Apollo Scientific
Warning

OR303186-1G OR303186-5G
APOSOR303186-1GEA 0 GBP
APOSOR303186-1G APOSOR303186-5G
tert-Butyl-3-hydroxypent-4-enoate
tert-Butyl-3-hydroxypent-4-enoate
Formula: C₉H₁₆O₃
Storage Temperature: Ambient
MDL Number: MFCD18207180
CAS Number: 122763-67-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR