N-Benzyl-N-methyl-β-alanine

Supplier: Apollo Scientific
Warning

Synonyms: 3-[Benzyl(methyl)amino]propanoic acid

OR303159-1G OR303159-5G
APOSOR303159-1GEA 129 GBP
APOSOR303159-1G APOSOR303159-5G
N-Benzyl-N-methyl-β-alanine
N-Benzyl-N-methyl-beta-alanine
Formula: C₁₁H₁₅NO₂
MW: 193,25 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD09717730
CAS Number: 149692-49-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR