3-Chloro-4-(4-methylpiperazin-1-yl)phenylamine

Supplier: Apollo Scientific

Synonyms: 3-Chloro-4-(4-methylpiperazin-1-yl)aniline, 3-Chloro-4-(4-methylpiperazin-1-yl)benzenamine

OR303155-1G OR303155-500MG
APOSOR303155-1GEA 295 GBP
APOSOR303155-1G APOSOR303155-500MG
3-Chloro-4-(4-methylpiperazin-1-yl)phenylamine
3-Chloro-4-(4-methylpiperazin-1-yl)phenylamine
Formula: C₁₁H₁₆ClN₃
MW: 225,72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01951505
CAS Number: 16154-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR