(2E)-2-{[4-Methoxy-3-(3-methoxypropoxy)phenyl]methylidene}-3-methylbutanoic acid

Supplier: Apollo Scientific

OR303035-1G OR303035-500MG
APOSOR303035-1GEA 453 GBP
APOSOR303035-1G APOSOR303035-500MG
(2E)-2-{[4-Methoxy-3-(3-methoxypropoxy)phenyl]methylidene}-3-methylbutanoic acid
(2E)-2-{[4-Methoxy-3-(3-methoxypropoxy)phenyl]methylidene}-3-methylbutanoic acid
Formula: C₁₇H₂₄O₅
Storage Temperature: Ambient
MDL Number: MFCD11112069
CAS Number: 387868-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR