(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid

Supplier: Apollo Scientific

Synonyms: (S)-2-(1-BOC-2-pyrrolidinyl)acetic acid, Boc-β-HomoPro-OH

OR302704-1G OR302704-5G
APOSOR302704-1GEA 85.2 GBP
APOSOR302704-1G APOSOR302704-5G
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid
(S)-2-(1-(tert-Butoxycarbonyl)pyrrolidin-2-yl)acetic acid
Formula: C₁₁H₁₉NO₄
MW: 229,3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01862939
CAS Number: 56502-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR