2,4,6-Trichloro-5-cyanopyrimidine

Supplier: Apollo Scientific
Warning

OR302358-1G OR302358-5G
APOSOR302358-1GEA 187 GBP
APOSOR302358-1G APOSOR302358-5G
2,4,6-Trichloro-5-cyanopyrimidine
2,4,6-Trichloro-5-cyanopyrimidine
Formula: C₅Cl₃N₃
MW: 208,4300 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07636745
CAS Number: 3029-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR