Methyl-2-cyanobenzoate

Supplier: Apollo Scientific
Warning

Synonyms: Methyl 2-cyanobenzoate, 2-Cyanobenzoic acid methyl ester

OR30234-10G OR30234-5G
APOSOR30234-10GEA 104 GBP
APOSOR30234-10G APOSOR30234-5G
Methyl-2-cyanobenzoate
Methyl 2-cyanobenzoate
Formula: C₉H₇NO₂
MW: 161,16 g/mol
Boiling Pt: 98…100 °C/0,2mmHg
Melting Pt: 47…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00016376
CAS Number: 6587-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR