2-Bromo-1-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethanone
Supplier: Apollo Scientific
|
Danger
|
OR30230-1G
OR30230-250MG
APOSOR30230-1GEA
0
GBP
APOSOR30230-1G
APOSOR30230-250MG
2-Bromo-1-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethanone
2-Bromo-1-(5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethanone
Formula:
C₁₆H₂₁BrO Melting Pt: 65…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00178768 CAS Number: 132392-28-0 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...