2-Amino-4-methylbenzonitrile
Supplier: Apollo Scientific
|
Warning
|
OR30201-1G
OR30201-5G
OR30201-25G
APOSOR30201-1GEA
19.7
GBP
APOSOR30201-1G
APOSOR30201-5G
APOSOR30201-25G
2-Amino-4-methylbenzonitrile
2-Amino-4-methylbenzonitrile
Formula:
C₈H₈N₂ Melting Pt: 91…93 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173706 CAS Number: 26830-96-6 EINECS: 248-020-0 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...