3-Amino-5-(2-furyl)-1H-pyrazole 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-(Fur-2-yl)-1H-pyrazol-3-amine, 5-(Furan-2-yl)-1H-pyrazol-3-amine
OR3019-1G
OR3019-250MG
APOSOR3019-1GEA
34.1
GBP
APOSOR3019-1G
APOSOR3019-250MG
3-Amino-5-(2-furyl)-1H-pyrazole 97%
3-Amino-5-(2-furyl)-1H-pyrazole
Formula:
C₇H₇N₃O Melting Pt: 124…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082696 CAS Number: 96799-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...