3,4-Dimethylbenzonitrile 98%
Supplier: Apollo Scientific
|
Warning
|
OR30142-10G
OR30142-25G
APOSOR30142-10GEA
61.7
GBP
APOSOR30142-10G
APOSOR30142-25G
3,4-Dimethylbenzonitrile 98%
3,4-Dimethylbenzonitrile
Formula:
C₉H₉N MW: 131,18 g/mol Boiling Pt: 230…232 °C Melting Pt: 64…68 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016380 CAS Number: 22884-95-3 EINECS: 245-293-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...